PloS one
Foxp1 regulates cortical radial migration and neuronal morphogenesis in developing cerebral cortex.
PMID 26010426
Abstract
FOXP1 is a member of FOXP subfamily transcription factors. Mutations in FOXP1 gene have been found in various development-related cognitive disorders. However, little is known about the etiology of these symptoms, and specifically the function of FOXP1 in neuronal development. Here, we report that suppression of Foxp1 expression in mouse cerebral cortex led to a neuronal migration defect, which was rescued by overexpression of Foxp1. Mice with Foxp1 knockdown exhibited ectopic neurons in deep layers of the cortex postnatally. The neuronal differentiation of Foxp1-downregulated cells was normal. However, morphological analysis showed that the neurons with Foxp1 deficiency had an inhibited axonal growth in vitro and a weakened transition from multipolar to bipolar in vivo. Moreover, we found that the expression of Foxp1 modulated the dendritic maturation of neurons at a late postnatal date. Our results demonstrate critical roles of Foxp1 in the radial migration and morphogenesis of cortical neurons during development. This study may shed light on the complex relationship between neuronal development and the related cognitive disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405681
Anti-CUX1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003277
Anti-CUX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003317
Anti-CUX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105137
Anti-CUX1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
AV30096
Anti-EOMES antibody produced in rabbit, affinity isolated antibody
HPA028896
Anti-EOMES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104422
Anti-EOMES antibody produced in rabbit, affinity isolated antibody
SAB2105420
Anti-EOMES antibody produced in rabbit, affinity isolated antibody
SAB2105298
Anti-EOMES, (C-terminal) antibody produced in rabbit, affinity isolated antibody
HPA003876
Anti-FOXP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32564
Anti-FOXP1 antibody produced in rabbit, IgG fraction of antiserum
SAB4503877
Anti-phospho-Histone H3 (pSer28) antibody produced in rabbit, affinity isolated antibody
SAB4503876
Anti-phospho-Histone H3 (pThr3) antibody produced in rabbit, affinity isolated antibody
PRS4627
Anti-SATB2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4629
Anti-SATB2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001042
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA029543
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102978
Anti-TBR1 antibody produced in rabbit, affinity isolated antibody
SAB2701324
Anti-TBR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2105287
Anti-TBR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0001523M1
Monoclonal Anti-CUTL1 antibody produced in mouse, clone 2A10, purified immunoglobulin, buffered aqueous solution
SAB1403715
Monoclonal Anti-CUTL1 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB1409861
Monoclonal Anti-EOMES antibody produced in mouse, clone 2D3, purified immunoglobulin, buffered aqueous solution
WH0027086M1
Monoclonal Anti-FOXP1 antibody produced in mouse, clone 4E3-G11, purified immunoglobulin, buffered aqueous solution
AMAB90682
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0323, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90680
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0321, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90679
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0320, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90678
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0319, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90635
Monoclonal Anti-SATB2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0276, purified immunoglobulin, buffered aqueous glycerol solution
WH0010716M1
Monoclonal Anti-TBR1 antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution