PloS one
Disabling Mitochondrial Peroxide Metabolism via Combinatorial Targeting of Peroxiredoxin 3 as an Effective Therapeutic Approach for Malignant Mesothelioma.
PMID 26011724
Abstract
Dysregulation of signaling pathways and energy metabolism in cancer cells enhances production of mitochondrial hydrogen peroxide that supports tumorigenesis through multiple mechanisms. To counteract the adverse effects of mitochondrial peroxide many solid tumor types up-regulate the mitochondrial thioredoxin reductase 2--thioredoxin 2 (TRX2)--peroxiredoxin 3 (PRX3) antioxidant network. Using malignant mesothelioma cells as a model, we show that thiostrepton (TS) irreversibly disables PRX3 via covalent crosslinking of peroxidatic and resolving cysteine residues in homodimers, and that targeting the oxidoreductase TRX2 with the triphenylmethane gentian violet (GV) potentiates adduction by increasing levels of disulfide-bonded PRX3 dimers. Due to the fact that activity of the PRX3 catalytic cycle dictates the rate of adduction by TS, immortalized and primary human mesothelial cells are significantly less sensitive to both compounds. Moreover, stable knockdown of PRX3 reduces mesothelioma cell proliferation and sensitivity to TS. Expression of catalase in shPRX3 mesothelioma cells restores defects in cell proliferation but not sensitivity to TS. In a SCID mouse xenograft model of human mesothelioma, administration of TS and GV together reduced tumor burden more effectively than either agent alone. Because increased production of mitochondrial hydrogen peroxide is a common phenotype of malignant cells, and TS and GV are well tolerated in mammals, we propose that targeting PRX3 is a feasible redox-dependent strategy for managing mesothelioma and other intractable human malignancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100850
Anti-FOXM1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100851
Anti-FOXM1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412254
ANTI-FOXM1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1412255
ANTI-FOXM1 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB1412256
ANTI-FOXM1 antibody produced in mouse, clone 3E3, purified immunoglobulin, buffered aqueous solution
SAB1412277
ANTI-FOXM1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
AV39518
Anti-FOXM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA029974
Anti-FOXM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA029976
Anti-FOXM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU114181 MISSION® esiRNA, esiRNA human PRDX3 (esiRNA1)
EMU040451 MISSION® esiRNA, esiRNA targeting mouse Prdx3 (esiRNA1)
SAB1403827
Monoclonal Anti-FOXM1, (C-terminal) antibody produced in mouse, clone 3A9, purified immunoglobulin, buffered aqueous solution
307823
Sodium hydrogen sulfate, technical grade
HNaO4S