EMAIL THIS PAGE TO A FRIEND
European journal of endocrinology
Prevalence and risk factors of gestational diabetes in Punjab, North India: results from a population screening program.
PMID 26012589
Abstract
The World Health Organization (WHO) has in 2013 changed the diagnostic criteria for gestational diabetes mellitus (GDM) to acknowledge the putative effect of mildly elevated fasting plasma glucose (FPG) levels on pregnancy outcomes. We aimed to determine the prevalence and risk factors of GDM comparing the previous WHO 1999 criteria to the WHO 2013 criteria in North India. In a population-based screening programme, 5100 randomly selected North Indian women were studied using a cross-sectional design with a questionnaire, venous FPG and 2-h capillary plasma glucose (PG) after a 75 g oral glucose tolerance test performed between 24 and 28 weeks of pregnancy. The prevalence of GDM was 35% using WHO 2013 criteria vs 9% using WHO 1999 criteria. FPG measurements identified 94% of WHO 2013 GDM cases as opposed to 11% of WHO 1999 GDM cases. In contrast, 2-h PG measurements identified only 13% of WHO 2013 GDM cases compared with 96% of the WHO 1999 GDM cases. Using logistic regression with backward elimination, urban habitat, illiteracy, non-vegetarianism, increased BMI, Hindu religion and low adult height were all independent risk factors of GDM using the 1999 criteria, whereas only urban habitat, low adult height and increased age were independent risk factors of GDM using the 2013 criteria. Intervention studies are needed to justify the WHO 2013 GDM criteria increasing the prevalence four fold to include more than one third of North Indian pregnant women.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50404
1,3-Propanediol, 98%
C3H8O2