EMAIL THIS PAGE TO A FRIEND
Stem cells (Dayton, Ohio)
The GIPC1-Akt1 Pathway Is Required for the Specification of the Eye Field in Mouse Embryonic Stem Cells.
PMID 26013465
Abstract
During early patterning of the neural plate, a single region of the embryonic forebrain, the eye field, becomes competent for eye development. The hallmark of eye field specification is the expression of the eye field transcription factors (EFTFs). Experiments in fish, amphibians, birds, and mammals have demonstrated largely conserved roles for the EFTFs. Although some of the key signaling events that direct the synchronized expression of these factors to the eye field have been elucidated in fish and frogs, it has been more difficult to study these mechanisms in mammalian embryos. In this study, we have used two different methods for directed differentiation of mouse embryonic stem cells (mESCs) to generate eye field cells and retina in vitro to test for a role of the PDZ domain-containing protein GIPC1 in the specification of the mammalian eye primordia. We find that the overexpression of a dominant-negative form of GIPC1 (dnGIPC1), as well as the downregulation of endogenous GIPC1, is sufficient to inhibit the development of eye field cells from mESCs. GIPC1 interacts directly with IGFR and participates in Akt1 activation, and pharmacological inhibition of Akt1 phosphorylation mimics the dnGIPC1 phenotype. Our data, together with previous studies in Xenopus, support the hypothesis that the GIPC1-PI3K-Akt1 pathway plays a key role in eye field specification in vertebrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22308F Anti-GIPC1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407046
Anti-GIPC1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701664
Anti-gipc1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV40601
Anti-GIPC1 antibody produced in rabbit, affinity isolated antibody
HPA043958
Anti-GIPC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500593
Anti-LHX2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100884
Anti-LHX2 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
AV38062
Anti-PAX6 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32741
Anti-PAX6 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406221
Anti-PAX6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA030775
Anti-PAX6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410879
Anti-PAX6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411780
ANTI-RCV1 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB1411781
ANTI-RCV1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1411794
ANTI-RCV1 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
SAB1411795
ANTI-RCV1 antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1411804
ANTI-RCV1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411805
ANTI-RCV1 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1411862
ANTI-RCV1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1411863
ANTI-RCV1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
SAB1411864
ANTI-RCV1 antibody produced in mouse, clone 4A8, purified immunoglobulin, buffered aqueous solution
SAB1411865
ANTI-RCV1 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
SAB1411878
ANTI-RCV1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1411879
ANTI-RCV1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
AV38196
Anti-RCV1 antibody produced in rabbit, affinity isolated antibody
SAB1406380
Anti-RCVRN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101971
Anti-RCVRN antibody produced in rabbit, affinity isolated antibody
SAB4200309
Anti-Recoverin (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB4502604
Anti-Recoverin antibody produced in rabbit, affinity isolated antibody
SAB5300261
Monoclonal Anti-LHX2 antibody produced in mouse, clone 6G2, ascites fluid