EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
Selective recruitment of nuclear factors to productively replicating herpes simplex virus genomes.
PMID 26018390
Abstract
Much of the HSV-1 life cycle is carried out in the cell nucleus, including the expression, replication, repair, and packaging of viral genomes. Viral proteins, as well as cellular factors, play essential roles in these processes. Isolation of proteins on nascent DNA (iPOND) was developed to label and purify cellular replication forks. We adapted aspects of this method to label viral genomes to both image, and purify replicating HSV-1 genomes for the identification of associated proteins. Many viral and cellular factors were enriched on viral genomes, including factors that mediate DNA replication, repair, chromatin remodeling, transcription, and RNA processing. As infection proceeded, packaging and structural components were enriched to a greater extent. Among the more abundant proteins that copurified with genomes were the viral transcription factor ICP4 and the replication protein ICP8. Furthermore, all seven viral replication proteins were enriched on viral genomes, along with cellular PCNA and topoisomerases, while other cellular replication proteins were not detected. The chromatin-remodeling complexes present on viral genomes included the INO80, SWI/SNF, NURD, and FACT complexes, which may prevent chromatinization of the genome. Consistent with this conclusion, histones were not readily recovered with purified viral genomes, and imaging studies revealed an underrepresentation of histones on viral genomes. RNA polymerase II, the mediator complex, TFIID, TFIIH, and several other transcriptional activators and repressors were also affinity purified with viral DNA. The presence of INO80, NURD, SWI/SNF, mediator, TFIID, and TFIIH components is consistent with previous studies in which these complexes copurified with ICP4. Therefore, ICP4 is likely involved in the recruitment of these key cellular chromatin remodeling and transcription factors to viral genomes. Taken together, iPOND is a valuable method for the study of viral genome dynamics during infection and provides a comprehensive view of how HSV-1 selectively utilizes cellular resources.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700474
Anti-ERCC3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2702179
Anti-ERCC3 antibody produced in rabbit
SAB2106419
Anti-ERCC3 antibody produced in rabbit, affinity isolated antibody
AV34116
Anti-G22P1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV34117
Anti-G22P1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405910
Anti-GTF2I antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV31910
Anti-GTF2I antibody produced in rabbit, affinity isolated antibody
HPA026638
Anti-GTF2I antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100991
Anti-GTF2I antibody produced in rabbit, affinity isolated antibody
SAB4300412
Anti-HDAC2 (Ab-394) antibody produced in rabbit, affinity isolated antibody
HPA011727
Anti-HDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32618
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB2104633
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB4501384
Anti-HDAC2, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV46122
Anti-HIST1H1C antibody produced in rabbit, affinity isolated antibody
SAB2701559
Anti-HIST1H1C antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
H3159
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H2663
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture
SAB4500408
Anti-Histone H1 (Acetyl-Lys25) antibody produced in rabbit, affinity isolated antibody
SAB4500409
Anti-Histone H1, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV38110
Anti-HMGB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2701685
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701809
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101049
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB4501401
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB1306517
ANTI-KU70 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
K4763 Anti-Ku70 antibody produced in rabbit, affinity isolated antibody, ammonium sulfate suspension
SAB4501057
Anti-KU70, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503096
Anti-KU70, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA002697
Anti-SSRP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution