EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Epigenetic changes as a common trigger of muscle weakness in congenital myopathies.
PMID 26019235
Abstract
Congenital myopathies are genetically and clinically heterogeneous conditions causing severe muscle weakness, and mutations in the ryanodine receptor gene (RYR1) represent the most frequent cause of these conditions. A common feature of diseases caused by recessive RYR1 mutations is a decrease of ryanodine receptor 1 protein content in muscle. The aim of the present investigation was to gain mechanistic insight into the causes of this reduced ryanodine receptor 1. We found that muscle biopsies of patients with recessive RYR1 mutations exhibit decreased expression of muscle-specific microRNAs, increased DNA methylation and increased expression of class II histone deacetylases. Transgenic mouse muscle fibres over-expressing HDAC-4/HDAC-5 exhibited decreased expression of RYR1 and of muscle-specific miRNAs, whereas acute knock-down of RYR1 in mouse muscle fibres by siRNA caused up-regulation of HDAC-4/HDAC-5. Intriguingly, increased class II HDAC expression and decreased ryanodine receptor protein and miRNAs expression were also observed in muscles of patients with nemaline myopathy, another congenital neuromuscular disorder. Our results indicate that a common pathophysiological pathway caused by epigenetic changes is activated in some forms of congenital neuromuscular disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300413
Anti-HDAC4 (Ab-632) antibody produced in rabbit, affinity isolated antibody
SAB2101022
Anti-HDAC4 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101023
Anti-HDAC4 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB4503697
Anti-HDAC4 antibody produced in rabbit, affinity isolated antibody
SAB1400126
Anti-HLA-E antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401182
Anti-HLA-E antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031454
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
A9917 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2304 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3682 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
A0168 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody
A2554 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9309 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3673 Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701131 Anti-Mouse IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701159 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
A8924 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A5278 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3701015 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700993 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
C0130
Collagenase from Clostridium histolyticum, for general use, Type I, 0.25-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
H3506
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
SAB4700300 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/08, purified immunoglobulin, buffered aqueous solution
SAB4700301 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/06, purified immunoglobulin, buffered aqueous solution
SAB4700302 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/07, purified immunoglobulin, buffered aqueous solution
P8170 Protein G−Peroxidase from Streptococcus sp., recombinant, expressed in unspecified host, lyophilized powder
SML1106
Ryanodine, ≥98% (HPLC)
C25H35NO9