EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Synthesis and antiproliferative evaluation of novel 5-(4-methylpiperazin-1-yl)-2-phenyl- 1H-benzimidazole derivatives.
PMID 26020557
Abstract
A series of novel 5-(4-methylpiperazin-1-yl)-2-phenyl-1H-benzimidazoles (5-14) were synthesized and evaluated for their in vitro antiproliferative activities against the human leukemia cell line HL-60. Compounds 5-7 and 10-12 exhibited potent antiproliferative activities against this cell line. The quantitative analysis of apoptosis by flow cytometry demonstrated that the percentages of apoptotic HL-60 cells treated with compounds 5 and 10-12 were significantly higher than in the control.