EMAIL THIS PAGE TO A FRIEND
Mutation research
Are effects of common ragwort in the Ames test caused by pyrrolizidine alkaloids?
PMID 26021695
Abstract
It has previously been demonstrated by others that acetone extracts of Senecio jacobaea (syn. Jacobaea vulgaris, common or tansy ragwort) test positive in the Salmonella/microsome mutagenicity test (Ames test). Pyrrolizidine alkaloids (PAs) are thought to be responsible for these mutagenic effects. However, it was also observed that the major PA present in common ragwort, jacobine, produced a negative response (with and without the addition of rat liver S9) in Salmonella test strains TA98, TA100, TA1535 and TA1537. To investigate which compounds in the plant extracts were responsible for the positive outcome, the present study investigated the contents and mutagenic effects of methanol and acetone extracts prepared from dried ground S. jacobaea and Senecio inaequidens (narrow-leafed ragwort). Subsequently, a fractionation approach was set up in combination with LC-MS/MS analysis of the fractions. It was shown that the positive Ames test outcomes of S. jacobaea extracts are unlikely to be caused by PAs, but rather by the flavonoid quercetin. This study also demonstrates the importance of identifying compounds responsible for positive test results in bioassays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A38800
2-Aminoanthracene, 96%
C14H11N
S9506
Ammonium sodium phosphate dibasic tetrahydrate, ≥99%
H5NNaO4P · 4H2O
S4172
Ammonium sodium phosphate dibasic tetrahydrate, BioXtra, ≥99%
H5NNaO4P · 4H2O
N5755
β-Nicotinamide adenine dinucleotide phosphate hydrate
C21H28N7O17P3 · xH2O
N8035
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
C21H27N7NaO17P3
N8160
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
C21H27N7NaO17P3
C1909
Citric acid monohydrate, ACS reagent, ≥99.0%
C6H8O7 · H2O
C7129
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
C6H8O7 · H2O
C0706
Citric acid monohydrate, BioXtra, ≥99.5%
C6H8O7 · H2O
16301
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 7.0-9.5% water(Karl Fischer)
C6H12O6 · H2O
D9559
Dextrose monohydrate, meets USP testing specifications
C6H12O6 · H2O
H6034
L-Histidine, cell culture tested, meets EP, USP testing specifications, from non-animal source
C6H9N3O2
H8000
L-Histidine, ReagentPlus®, ≥99% (TLC)
C6H9N3O2
H3911
L-Histidine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H9N3O2
53319
L-Histidine, BioUltra, ≥99.5% (NT)
C6H9N3O2
63138
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
MgO4S · 7H2O
RES0089M-A7
Magnesium sulfate heptahydrate, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
MgO4S · 7H2O
M5921
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
MgO4S · 7H2O
M2773
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
MgO4S · 7H2O
R0382
Retrorsine, ≥90% (HPLC)
C18H25NO6
17806
Senecionine, ≥95.0% (GC)
C18H25NO5
71643
Sodium phosphate dibasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
HNa2O4P · 2H2O
RES0974S-A7
Sodium phosphate dibasic dihydrate, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
HNa2O4P · 2H2O