EMAIL THIS PAGE TO A FRIEND
Scientific reports
Decreased expression of CHIP leads to increased angiogenesis via VEGF-VEGFR2 pathway and poor prognosis in human renal cell carcinoma.
PMID 26021863
Abstract
CHIP (c-terminal Hsp70-interacting protein) is an E3 ligase which may play different roles in different cancers. The elucidation of the VHL-HIF-1α (hypoxia inducible factor-1α)-VEGF (vascular endothelial growth factor) pathway has led to the development of targeted therapy in renal cell carcinoma (RCC). However, little is known about the role of CHIP and the relationship between CHIP and VEGF-VEGFR2 (VEGF receptor 2) pathway in RCC. In this study, we found that the expression of CHIP was downregulated and significantly correlated with pT status (P = 0.022) and TNM stage (P = 0.022) in 304 RCC and 35 normal renal tissues using tissue microarray. Moreover, low expression of CHIP is a strong and independent negative prognostic value for RCC. In vitro, CHIP negatively regulated RCC cell migration, invasion and angiogenesis. In addition, ELISA tests showed that restoration of CHIP inhibited, while knockdown promoted, the secreted level of VEGF. Furthermore, western blot indicated that the VEGFR2 protein level was reduced after CHIP overexpression. Our findings demonstrate for the first time that CHIP may be involved in RCC angiogenesis through regulating VEGF secretion and expression of VEGFR2. CHIP may serve as promising prognostic biomarker of angiogenesis and may constitute a potential therapeutic target in RCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9243
Anti-CHIP (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
C9118
Anti-CHIP (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
C5617 Anti-CHIP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2102328
Anti-STUB1 antibody produced in rabbit, affinity isolated antibody
HPA041222
Anti-STUB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA043531
Anti-STUB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103799
Anti-STUB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4501642
Anti-VEGFR2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501643
Anti-VEGFR2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501644
Anti-VEGFR2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500965
Anti-VEGFR2, C-Terminal antibody produced in rabbit, affinity isolated antibody
K2643 KDR (789-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
EHU106761 MISSION® esiRNA, esiRNA human STUB1 (esiRNA1)
EMU091541 MISSION® esiRNA, esiRNA targeting mouse Stub1 (esiRNA1)
SAB1403057
Monoclonal Anti-STUB1, (C-terminal) antibody produced in mouse, clone 2E12, purified immunoglobulin, buffered aqueous solution