EMAIL THIS PAGE TO A FRIEND
Diabetes research and clinical practice
High glucose and/or high insulin affects HIF-1 signaling by regulating AIP1 in human umbilical vein endothelial cells.
PMID 26021979
Abstract
The objective of this study was to explore the effects of high glucose/high insulin on AIP1 expression in HUVECs and the possible regulation of HIF-1α signaling by AIP1. We investigated the expression of AIP1 and HIF-1α signaling in HUVECs at the levels of mRNA and protein following exposure to 30 mmol/L glucose (high glucose), 1 nmol/L insulin (high insulin), and the combination of the two (high glucose/high insulin). We detected changes in HIF-1α and VEGF expression with AIP1 siRNA interference by real-time PCR and western blotting. The CCK8 cell proliferation assay, the scratch/wound-healing assay, and flow cytometry were used to assess cell proliferation, migration and apoptosis, respectively. Matrigel was used to perform a tubule formation assay. Compared with 5.5 mmol/L glucose alone (control), high glucose, high insulin, and the combination of high glucose+high insulin increased AIP1 expression at 24 h at the mRNA and protein levels. High glucose, high insulin, and high glucose+high insulin decreased HIF-1α expression at the mRNA and protein levels. AIP1 knockdown significantly increased HIF-1α and VEGF expression at both the mRNA and protein levels in HUVECs under high glucose conditions. In the presence of high insulin, the effect of high glucose on target gene expression was altered. The downregulation of AIP1 promoted cell proliferation, migration, and tubule formation, and it decreased apoptosis. High glucose increases AIP1 expression and decreases the expression of HIF-1α and downstream molecules. Decreased HIF-1α signaling may be regulated by increased AIP1 under high glucose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1408334
Anti-ACTRT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003119 Anti-ACTRT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103464
Anti-ACTRT1 antibody produced in rabbit, affinity isolated antibody
SAB4200477
Anti-ALIX (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200476
Anti-ALIX antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1400447
Anti-ARL6IP1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA045307
Anti-ARL6IP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3305
Anti-ARMER antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900355
Anti-ARMER antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501559
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501560
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1409216
Anti-BIRC3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002317
Anti-BIRC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401013
Anti-BIRC3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA036977
Anti-DAB2IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M2441 Anti-MAGI-2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA013650
Anti-MAGI2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503718
Anti-MAGI2 antibody produced in rabbit, affinity isolated antibody
SAB1406912
Anti-PDCD6IP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011905
Anti-PDCD6IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410056
Anti-PDCD6IP antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV41301
Anti-WDR1 antibody produced in rabbit, affinity isolated antibody
SAB2700801
Anti-WDR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700803
Anti-WDR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA036434
Anti-WDR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105449
Anti-WDR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1403128 Monoclonal Anti-ARL6IP antibody produced in mouse, clone 4F9, purified immunoglobulin, buffered aqueous solution
SAB4100427
Monoclonal Anti-BIRC3 antibody produced in mouse, clone BIRC3-A359, culture supernatant
WH0009863M1
Monoclonal Anti-MAGI2 antibody produced in mouse, clone 6C8, purified immunoglobulin, buffered aqueous solution
SAB1404762
Monoclonal Anti-MAGI2 antibody produced in mouse, clone 6F5, purified immunoglobulin, buffered aqueous solution