EMAIL THIS PAGE TO A FRIEND
Biomaterials
Low-aspect ratio nanopatterns on bioinert alumina influence the response and morphology of osteoblast-like cells.
PMID 26022980
Abstract
Topographical features on the nanometer scale are known to influence cellular behavior. The response of specific cell types to various types of surface structures is currently still being investigated. Alumina ceramics play an important role as biomaterials, e.g., in medical and dental applications. In this study, we investigated the influence of nanoscale surface features with low aspect ratio (< 0.1) on the response of osteoblast-like MG-63 cells. To this end, low-energy ion irradiation was employed to produce shallow nanoscale ripple patterns on Al2O3(0001) surfaces with lateral periodicities of 24 nm and 179 nm and heights of only 0.7 and 11.5 nm, respectively. The nanopatterning was found to increase the proliferation of MG-63 cells and may lead to pseudopodia alignment along the ripples. Furthermore, focal adhesion behavior and cell morphology were analyzed. We found that MG-63 cells are able to recognize surface nanopatterns with extremely low vertical variations of less than 1 nm. In conclusion, it is shown that surface topography in the sub-nm range significantly influences the response of osteoblast-like cells.