EMAIL THIS PAGE TO A FRIEND
Journal of toxicologic pathology
Establishment of a syngeneic orthotopic model of prostate cancer in immunocompetent rats.
PMID 26023257
Abstract
We previously established 3 cell lines (PLS10, PLS20 and PLS30) from a chemically-induced prostate carcinoma in F344 rats, and demonstrated high potential for metastasis in nude mice. In the present study, we investigated the feasibility of establishing an orthotopic model using the 3 rat prostate cancer cell lines in immunocompetent rats with the aim of resolving species-mismatch problems and defects of immune systems. The PLS10, PLS20 and PLS30 cell lines were injected into the ventral prostates of 6-week-old rats, which were then sacrificed at experimental weeks 4 and 8. Tumor mass formation was found in rats with PLS10, but not in those with PLS20 or PLS30. Additionally, metastatic carcinomas could be detected in lymph nodes and lungs of PLS10-inoculated rats. Genetic analysis demonstrated K-ras gene mutations in PLS10 and PLS20, but not in PLS30 cells. There were no mutations in p53 and KLF6. In conclusion, we established a syngeneic orthotopic model for prostate cancer in immunocompetent rats simulating human castration-resistant prostate cancer (CRPC), which should prove useful for development and validation of therapeutic agents, especially with immunotherapy.