EMAIL THIS PAGE TO A FRIEND
Cancer immunology research
STAT3 in CD8+ T Cells Inhibits Their Tumor Accumulation by Downregulating CXCR3/CXCL10 Axis.
PMID 26025380
Abstract
One of the obstacles for cancer immunotherapy is the inefficiency of CD8(+) T-cell recruitment to tumors. STAT3 has been shown to suppress CD8(+) T-cell antitumor functions in various cancer models, in part by restricting accumulation of CD8(+) T cells. However, the underlying molecular mechanism by which STAT3 in CD8(+) T cells inhibits their accumulation in tumors remains to be defined. Here, we show that STAT3 signaling in CD8(+) T cells inhibits chemokine CXCL10 production by tumor-associated myeloid cells by reducing IFNγ expression by T cells. We further demonstrate that ablating STAT3 in T cells allows expression of CXCR3, the receptor of CXCL10, on CD8(+) T cells, resulting in efficient accumulation of CD8(+) T cells at tumor sites. Blocking IFNγ or CXCR3 impairs the accumulation of STAT3-deficient CD8(+) T cells in tumor and their antitumor effects. Together, our study reveals a negative regulation by STAT3 signaling in T cells on cross-talk between myeloid cells and T cells through IFNγ/CXCR3/CXCL10, which is important for CD8(+) T cells homing to tumors. Our results thus provide new insights applicable to cancer immunotherapy and adoptive T-cell strategies.