EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Tulane Virus as a Potential Surrogate To Mimic Norovirus Behavior in Oysters.
PMID 26025893
Abstract
Oyster contamination by noroviruses is an important health and economic problem. The present study aimed to compare the behaviors of Norwalk virus (the prototype genogroup I norovirus) and two culturable viruses: Tulane virus and mengovirus. After bioaccumulation, tissue distributions were quite similar for Norwalk virus and Tulane virus, with the majority of viral particles detected in digestive tissues, while mengovirus was detected in large amounts in the gills and mantle as well as in digestive tissues. The levels of persistence of all three viruses over 8 days were comparable, but clear differences were observed over longer periods, with Norwalk and Tulane viruses displaying rather similar half-lives, unlike mengovirus, which was cleared more rapidly. These results indicate that Tulane virus may be a good surrogate for studying norovirus behavior in oysters, and they confirm the prolonged persistence of Norwalk virus in oyster tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0138
Tetrazolium Violet
C23H17ClN4