EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
miR-27 is associated with chemoresistance in esophageal cancer through transformation of normal fibroblasts to cancer-associated fibroblasts.
PMID 26026166
Abstract
There is increasing evidence that the expression of microRNA (miRNA) in cancer is associated with chemosensitivity but the mechanism of miRNA-induced chemoresistance has not been fully elucidated. The aim of this study was to examine the role of extracellular miRNA in the response to chemotherapy in esophageal cancer. First, serum expression of miRNAs selected by miRNA array was measured by quantitative reverse transcription-polymerase chain reaction in 68 patients with esophageal cancer who received cisplatin-based chemotherapy to examine the relationship between miRNA expression and response to chemotherapy. The serum expression levels of 18 miRNAs were different between responders and non-responders by miRNA array. Of these, high expression levels of miR-27a/b correlated with poor response to chemotherapy in patients with esophageal cancer. Next, in vitro assays were conducted to investigate the mechanism of miRNA-induced chemoresistance. Although transfection of miR-27a/b to cancer cells had no significant impact on chemosensitivity, esophageal cancer cells cultured in supernatant of miR-27a/b-transfected normal fibroblast showed reduced chemosensitivity to cisplatin, compared with cancer cells cultured in supernatant of normal fibroblast. MiR-27a/b-transfected normal fibroblast showed α-smooth muscle actin (α-SMA) expression, a marker of cancer-associated fibroblasts (CAF) and increased production of transforming growth factor-β (TGF-β). Chemosensitivity recovered after administration of neutralizing antibody of TGF-β to the supernatant transfer experiments. Our results indicated that miR-27a/b is involved in resistance to chemotherapy in esophageal cancer, through miR-27a/b-induced transformation of normal fibroblast into CAF.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21421 Anti-MITF antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA003259
Anti-MITF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37977
Anti-MITF antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2101480
Anti-MITF antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105416
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB4501879
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
479306
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
H6Cl2N2Pt
P4394
cis-Diammineplatinum(II) dichloride, crystalline
H6Cl2N2Pt
RAB0306
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0307
Human IL-6 ELISA Kit, for cell and tissue lysates
RAB0313
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
P248 Monoclonal Anti-PARP antibody produced in mouse, clone C-2-10, ascites fluid
WH0000142M1
Monoclonal Anti-PARP1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
SAB1403527
Monoclonal Anti-PARP1, (N-terminal) antibody produced in mouse, clone 2C7, purified immunoglobulin, buffered aqueous solution
RAB0308
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0309
Mouse IL-6 ELISA Kit, for cell and tissue lysates
RAB0314 Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
PLA0184
Rabbit anti-PARP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
RAB0311
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
RAB0312
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
P1525
trans-Platinum(II)diammine dichloride
H6Cl2N2Pt