EMAIL THIS PAGE TO A FRIEND
Metabolism: clinical and experimental
Splenectomy attenuates obesity and decreases insulin hypersecretion in hypothalamic obese rats.
PMID 26026366
Abstract
Obesity-induced abnormalities, such as insulin resistance, dyslipidemia and hypertension, are frequently correlated with low-grade inflammation, a process that may depend on normal spleen function. This study investigated the role of the spleen in the obesity induced by monosodium glutamate (MSG) treatment. MSG-obese and lean control (CON) rats were subjected to splenectomy (SPL) or non-operated (NO). MSG-NO rats presented a high adipose tissue content, insulin resistance, dyslipidemia and islet hypersecretion, accompanied by hypertrophy of both pancreatic islets and adipocytes when compared with CON-NO rats. In addition, changes in nitric oxide response were found in islets from the MSG-NO group without associated alterations in inducible nitric oxide synthase (iNOS) or IL1β expression. MSG-NO also presented increased leukocyte counts and augmented LPS-induced nitric oxide production in macrophages. Splenectomy of MSG-obese animals decreased insulin hypersecretion, normalized the nitric oxide response in the pancreatic islets, improved insulin sensitivity and reduced hypertrophy of both adipocytes and islets, when compared with MSG-NO rats. Results show that splenectomy attenuates the progression of the obesity modulating pancreas functions in MSG-obese rats.