EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Biocompatibility of Textile Titanium Nickel Implants with Fibroblast Culture.
PMID 26028231
Abstract
The parameters of biocompatibility of titanium nickel implants of different design with fibroblast culture are studied. Colonization of textile and mesh implants with fibroblasts and tissue development depend on the size of mesh cells and thread diameter. Titanium nickel implants of different constructions do not inhibit the growth of fibroblast culture.