EMAIL THIS PAGE TO A FRIEND
Anaerobe
In vitro modulation of probiotic bacteria on the biofilm of Candida glabrata.
PMID 26028405
Abstract
A conspicuous new concept of pathogens living as the microbial societies in the human host rather than free planktonic cells has raised considerable concerns among scientists and clinicians. Fungal biofilms are communities of cells that possess distinct characteristic such as increased resistance to the immune defence and antimycotic agents in comparison to their planktonic cells counterpart. Therefore, inhibition of the biofilm may represent a new paradigm for antifungal development. In this study, we aim to evaluate the inxa0vitro modulation of vulvovaginal candidiasis (VVC)-causing Candida glabrata biofilms using probiotic lactobacilli strains. Probiotic Lactobacillus rhamnosus GR-1 and Lactobacillus reuteri RC-14 were shown to have completely inhibited C. glabrata biofilms and the results were corroborated by scanning electron microscopy (SEM), which revealed scanty structures of the mixed biofilms of C.xa0glabrata and probiotic lactobacilli strains. In addition, biofilm-related C.xa0glabrata genes EPA6 and YAK1 were downregulated in response to the probiotic lactobacilli challenges. The present study suggested that probiotic L.xa0rhamnosus GR-1 and L.xa0reuteri RC-14 strains inhibited C.xa0glabrata biofilm by partially impeding the adherence of yeast cells and the effect might be contributed by the secretory compounds produced by these probiotic lactobacilli strains. Further investigations are required to examine and identify the biofilm inhibitory compounds and the mechanism of probiotic actions of these lactobacilli strains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X4626
XTT sodium salt, powder, BioReagent, suitable for cell culture
C22H16N7NaO13S2