EMAIL THIS PAGE TO A FRIEND
Journal of Korean medical science
Treatment Response Evaluation of Breast Cancer after Neoadjuvant Chemotherapy and Usefulness of the Imaging Parameters of MRI and PET/CT.
PMID 26028936
Abstract
This study was aimed to evaluate the ability of imaging parameters measured on dynamic contrast-enhanced magnetic resonance imaging (DCE-MRI), diffusion-weighted MRI (DWI) and positron emission tomography/computed tomography (PET/CT) to serve as response markers in breast cancer after neoadjuvant chemotherapy (NAC). In 20 patients with breast cancer, DCE-MRI and DWI using a 3 T scanner and PET/CT were performed before and after NAC. DCE-MRI was analyzed using an automatic computer-aided detection program (MR-CAD). The response imaging parameters were compared with the pathologic response. The areas under the curve (AUCs) for DCE-MRI using MR-CAD analysis, DWI and PET/CT were 0.77, 0.59 and 0.76, respectively. The combination of all parameters measured by MR-CAD showed the highest diagnostic performance and accuracy (AUC = 0.77, accuracy = 90%). The combined use of the parameters of PET/CT with DCE-MRI or DWI showed a trend toward improved specificity and negative predictive value (100%, 100%, accuracy = 87.5%). The use of DCE-MRI using MR-CAD parameters indicated better diagnostic performance in predicting the final pathological response compared with DWI and PET/CT, although no statistically significant difference was observed. The combined use of PET/CT with DCE-MRI or DWI may improve the specificity for predicting a pathological response.