EMAIL THIS PAGE TO A FRIEND
Nature genetics
Mutations in the unfolded protein response regulator ATF6 cause the cone dysfunction disorder achromatopsia.
PMID 26029869
Abstract
Achromatopsia (ACHM) is an autosomal recessive disorder characterized by color blindness, photophobia, nystagmus and severely reduced visual acuity. Using homozygosity mapping and whole-exome and candidate gene sequencing, we identified ten families carrying six homozygous and two compound-heterozygous mutations in the ATF6 gene (encoding activating transcription factor 6A), a key regulator of the unfolded protein response (UPR) and cellular endoplasmic reticulum (ER) homeostasis. Patients had evidence of foveal hypoplasia and disruption of the cone photoreceptor layer. The ACHM-associated ATF6 mutations attenuate ATF6 transcriptional activity in response to ER stress. Atf6(-/-) mice have normal retinal morphology and function at a young age but develop rod and cone dysfunction with increasing age. This new ACHM-related gene suggests a crucial and unexpected role for ATF6A in human foveal development and cone function and adds to the list of genes that, despite ubiquitous expression, when mutated can result in an isolated retinal photoreceptor phenotype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5407
Anti-Opsin Antibody, blue, 100 µg, This Anti-Opsin Antibody, blue is validated for use in IH(P) for the detection of Opsin.
AB5405
Anti-Opsin Antibody, Red/Green, 100 µg, Anti-Opsin Antibody, Red/Green detects level of Opsin & has been published & validated for use in IH(P).
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501845
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501851
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003230
Anti-PDIA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution