EMAIL THIS PAGE TO A FRIEND
Vox sanguinis
Efficacy of the Mirasol pathogen reduction technology system against severe fever with thrombocytopenia syndrome virus (SFTSV).
PMID 26031768
Abstract
Severe fever with thrombocytopenia syndrome virus (SFTSV) is a tickborne virus in the Bunyaviridae family. This virus has recently been found in China, Japan and Korea. The risk of transfusion-transmitted SFTSV infection (TTI-SFTSV) is a concern because person-to-person transmission resulting from contact with SFTSV-contaminated blood has been reported. Therefore, we investigated the efficacy of the Mirasol pathogen reduction technology (PRT) system for inactivating SFTSV in vitro. The Mirasol PRT system achieved a > 4.11 log10 reduction value (LRV) for SFTSV. In conclusion, we showed that the Mirasol PRT system could potentially be used to reduce the risk of TTI-SFTSV.