EMAIL THIS PAGE TO A FRIEND
Journal of anatomy
Dynamic expression patterns of Pax6 during spermatogenesis in the mouse.
PMID 26032914
Abstract
Spermatogenesis is a series of complex processes to generate mature sperm, and various molecules play crucial roles in regulating these processes. Previous studies imply a possibility that a transcriptional factor Pax6, a key player of brain and sensory organ development, could be involved in spermatogenesis, but neither expression nor function of Pax6 in the adult testis has been examined yet. In the present study, we described for the first time Pax6 expression dynamics in the adult mouse testis. Using cell-type-specific markers, the expression of Pax6 was detected in 67.0% of promyelocytic leukemia zinc finger (Plzf)-positive type A spermatogonia. The expression of Pax6 was also observed in p63-positive spermatocytes and round spermatids. We did not detect any expression of Pax6 in Sox9-positive Sertoli cells or in elongated spermatids and mature sperm. High-resolution analyses revealed that Pax6 formed a single dot-like structure during mid-phase of the pachytene spermatocyte. This dot-like structure co-localized with γH2A.X demarcating XY body, a domain in which X and Y chromosomes are silenced and compartmentalized. These results may suggest a novel role of Pax6 in spermatogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1303846
ANTI-CTDSPL (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV42261
Anti-CTDSPL antibody produced in rabbit, affinity isolated antibody
HPA048325
Anti-CTDSPL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
SAB4501369
Anti-Histone H2AX , C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1306016
ANTI-PLZF(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1412499
ANTI-SOX9 antibody produced in mouse, clone 2A2, purified immunoglobulin, buffered aqueous solution
HPA001758
Anti-SOX9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37986
Anti-SOX9 antibody produced in rabbit, affinity isolated antibody
AV100797
Anti-SOX9 antibody produced in rabbit, IgG fraction of antiserum
SAB4502834
Anti-SOX9 antibody produced in rabbit, affinity isolated antibody
SAB2102346
Anti-SYCP3 antibody produced in rabbit, affinity isolated antibody
SAB1401670
Anti-SYCP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA043938
Anti-SYCP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039635
Anti-SYCP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501026
Anti-TCP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA027337
Anti-TCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA031082
Anti-TCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB2701838
Anti-tp63 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104972
Anti-TP63, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2502059
Anti-Trp63 (C OR N-TERMINAL) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001499
Anti-ZBTB16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PLA0294
Goat anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0003014M5 Monoclonal Anti-H2AFX antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
SAB1403902 Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
WH0006662M2
Monoclonal Anti-SOX9 antibody produced in mouse, clone 3C10, purified immunoglobulin, buffered aqueous solution
AMAB90795
Monoclonal Anti-SOX9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0639, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300368
Monoclonal Anti-SOX9 antibody produced in mouse, clone 1B11, purified immunoglobulin, buffered aqueous solution
SAB1404394
Monoclonal Anti-SOX9, (C-terminal) antibody produced in mouse, clone 3F11, purified immunoglobulin, buffered aqueous solution
WH0006950M1
Monoclonal Anti-TCP1 antibody produced in mouse, clone 2B2-D6, purified immunoglobulin, buffered aqueous solution