EMAIL THIS PAGE TO A FRIEND
Biology of the cell
Possible role of cortactin phosphorylation by protein kinase Cα in actin-bundle formation at growth cone.
PMID 26033110
Abstract
Cortactin contributes to growth cone morphogenesis by forming with dynamin, ring-shaped complexes that mechanically bundle and stabilise F-actin. However, the regulatory mechanism of cortactin action is poorly understood. Immunofluorescence microscopy revealed that protein kinase C (PKC) α colocalises with cortactin at growth cone filopodia in SH-SY5Y neuroblastoma cells. PKC activation by phorbol 12-myristate 13-acetate causes cortactin phosphorylation, filopodial retraction and F-actin-bundle loss. Moreover, PKCα directly phosphorylates cortactin in vitro at S135/T145/S172, mitigating both cortactin's actin-binding and actin-crosslinking activity, whereas cellular expression of a phosphorylation-mimetic cortactin mutant hinders filopodial formation with a significant decrease of actin bundles. Our results indicate that PKC-mediated cortactin phosphorylation might be implicated in the maintenance of growth cone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6987 Anti-Cortactin (GK-18) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
C7112 Anti-Cortactin (KE-20) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500765
Anti-Cortactin antibody produced in rabbit, affinity isolated antibody
SAB4500766
Anti-Cortactin, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300517
Anti-CTTN (Ab-421) antibody produced in rabbit, affinity isolated antibody
SAB4300518
Anti-CTTN (Ab-466) antibody produced in rabbit, affinity isolated antibody
AV52067
Anti-CTTN antibody produced in rabbit, affinity isolated antibody
SAB3701020 Anti-Mouse IgG (Fc specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701034 Anti-Mouse IgG (γ-chain specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701162 Anti-Mouse IgG (H+L), F(ab) fragment-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701113 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3701099 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3701077 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701092 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in sheep, affinity isolated antibody, lyophilized powder
SAB3700998 Anti-Mouse IgG F(ab′)2, highly cross adsorbed-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700997 Anti-Mouse IgG F(ab′)2-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1300371
Anti-PKCγ (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB5300316 Monoclonal Anti-CTTN antibody produced in mouse, clone 4C6, ascites fluid
SAB1402182
Monoclonal Anti-CTTN antibody produced in mouse, clone 2B5, purified immunoglobulin, buffered aqueous solution