EMAIL THIS PAGE TO A FRIEND
Biotechnology progress
Pretreatment of coconut mill effluent using celite-immobilized hydrolytic enzyme preparation from Staphylococcus pasteuri and its impact on anaerobic digestion.
PMID 26033963
Abstract
Biological treatment of oil and grease (O&G)-containing industrial effluents has long been a challenging issue. Practically feasible avenues to bring down their O&G load and enhance treatability are desired. In one such endeavour, the partially purified lipase from Staphylococcus pasteuri COM-4A was immobilized on celite carrier and applied for the enzymatic hydrolysis of unsterilized coconut oil mill effluent. In batch hydrolysis experiments, optimum conditions of 1% (w/v) immobilized lipase beads, one in four effluent dilution, and a contact time of 30 h resulted in 46% and 24% increase in volatile fatty acids and long-chain fatty acids and a concomitant 52% and 32% decrease in O&G and chemical oxygen demand (COD) levels, respectively. Batch anaerobic biodegradation trials with this prehydrolyzed effluent showed 89%, 91%, and 90% decrease in COD, proteins, and reducing sugars, respectively. These results were validated in a hybrid stirred tank--upflow anaerobic sludge blanket reactor. Average COD and O&G reductions effected by the hybrid reactor were found to be 89% and 88%, whereas that by the control reactor without enzymatic hydrolysis were only 60% and 47%, respectively. A maximum of 0.86 L methane gas was generated by the hybrid reactor per gram of VS added. Hence, this celite-immobilized crude lipase, sourced from a native laboratory isolate, seems to be a workable alternative to commercial enzyme preparations for the management of lipid-rich industrial effluents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N2752
4-Nitrophenyl palmitate, lipase substrate
C22H35NO4