EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Functionalized Collagen Scaffold Neutralizing the Myelin-Inhibitory Molecules Promoted Neurites Outgrowth in Vitro and Facilitated Spinal Cord Regeneration in Vivo.
PMID 26034998
Abstract
Research has demonstrated that many myelin-associated inhibitory molecules jointly contribute to the failure of adult spinal cord regeneration. Therapies comprehensively targeting the multiple inhibitory nature of the injured spinal cord are being concerned. Here, two collagen-binding proteins, CBD-EphA4LBD and CBD-PlexinB1LBD, were constructed, respectively, to neutralize the axon guidance molecules ephrinB3 and sema4D that inhibit the regeneration of nerve fibers. The two neutralizing proteins have proven their ability to specifically bind collagen and to continuously release from collagen scaffolds. They could also promote neurites outgrowth of cerebellar granular neurons and dorsal root ganglion neurons in vitro. Subsequently, the functionalized collagen scaffolds by physically absorbing NEP1-40 and immobilizing CBD-EphA4LBD and CBD-PlexinB1LBD were transplanted into a rat T10 complete spinal cord transection model. Our results showed that rats that received the treatment of transplanting the functionalized collagen scaffold exhibited great advantage on axonal regeneration and locomotion recovery after spinal cord injury.