EMAIL THIS PAGE TO A FRIEND
Pancreas
Mucin 1 Regulates Cox-2 Gene in Pancreatic Cancer.
PMID 26035123
Abstract
Eighty percent of pancreatic ductal adenocarcinomas (PDAs) overexpress mucin 1 (MUC1), a transmembrane mucin glycoprotein. MUC1(high) PDA patients also express high levels of cyclooxygenase 2 (COX-2) and show poor prognosis. The cytoplasmic tail of MUC1 (MUC1-CT) partakes in oncogenic signaling, resulting in accelerated cancer progression. Our aim was to understand the regulation of Cox-2 expression by MUC1. Levels of COX-2 and MUC1 were determined in MUC1(-/-), MUC1(low), and MUC1(high) PDA cells and tumors using reverse transcriptase-polymerase chain reaction, Western blot, and immunohistochemistry. Proliferative and invasive potential was assessed using MTT and Boyden chamber assays. Chromatin immunoprecipitation was performed to evaluate binding of MUC1-CT to the promoter of COX-2 gene. Significantly higher levels of COX-2 mRNA and protein were detected in MUC1(high) versus MUC1(low/null) cells, which were recapitulated in vivo. In addition, deletion of MUC1 gene and transient knockdown of MUC1 led to decreased COX-2 level. Also, MUC1-CT associated with the COX-2 promoter at ∼1000 base pairs upstream of the transcription start site, the same gene locus where nuclear factor κB p65 associates with the COX-2 promoter. Data supports a novel regulation of COX-2 gene by MUC1 in PDA, the intervention of which may lead to a better therapeutic targeting in PDA patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU095251 MISSION® esiRNA, esiRNA human MUC1 (esiRNA1)