EMAIL THIS PAGE TO A FRIEND
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment
Influence of the interactions between tea (Camellia sinensis) extracts and ascorbic acid on their antioxidant activity: analysis with interaction indexes and isobolograms.
PMID 26035225
Abstract
Products containing natural additives, including antioxidants, are usually perceived by consumers as safer than those with synthetic ones. Natural antioxidants, besides having a preservative activity, may exert beneficial health effects. Interactions between antioxidants may significantly change their antioxidant activity, thus in designing functional foods or food/cosmetic ingredients knowledge about the type of interactions could be useful. In the present study, the interactions between ascorbic acid (AA; vitamin C) and different black and green tea extracts and the influence on their antioxidant activities were investigated. The antioxidant activities of tea extracts and their mixtures with AA prepared in several different weight ratios were measured using the trolox equivalent antioxidant capacity (TEAC), 1,1-diphenyl-2-picrylhydrazyl (DPPH), and ferric-reducing antioxidant power (FRAP) methods. The type of interaction was determined by interaction indexes and isobolograms. It was found that the weight ratio of extracts to AA significantly influenced the antioxidant activity of a mixture and the type of interaction between these components. The weight ratio of tea extract to AA can cause the change of interaction, e.g. from antagonism to additivism or from additivism to synergism. The observed differences in the type of interactions were probably also a result of different extracts' polyphenol composition and content. The type of interaction may also be affected by the medium in which extracts and AA interact, especially its pH and the solvent used. To obtain the best antioxidant effect, all these factors should be taken into account during the design of a tea extract-AA mixture.