EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Effect of oral taurine on morbidity and mortality in elderly hip fracture patients: a randomized trial.
PMID 26035756
Abstract
Hip fracture patients represent a large part of the elderly surgical population and face severe postoperative morbidity and excessive mortality compared to adult surgical hip fracture patients. Low antioxidant status and taurine deficiency is common in the elderly, and may negatively affect postoperative outcome. We hypothesized that taurine, an antioxidant, could improve clinical outcome in the elderly hip fracture patient. A double blind randomized, placebo controlled, clinical trial was conducted on elderly hip fracture patients. Supplementation started after admission and before surgery up to the sixth postoperative day. Markers of oxidative status were measured during hospitalization, and postoperative outcome was monitored for one year after surgery. Taurine supplementation did not improve in-hospital morbidity, medical comorbidities during the first year, or mortality during the first year. Taurine supplementation lowered postoperative oxidative stress, as shown by lower urinary 8-hydroxy-2-deoxyguanosine levels (Generalized estimating equations (GEE) analysis average difference over time; regression coefficient (Beta): -0.54; 95% CI: -1.08--0.01; p = 0.04), blunted plasma malondialdehyde response (Beta: 1.58; 95% CI: 0.00-3.15; p = 0.05) and a trend towards lower lactate to pyruvate ratio (Beta: -1.10; 95% CI: -2.33-0.12; p = 0.08). We concluded that peri-operative taurine supplementation attenuated postoperative oxidative stress in elderly hip fracture patients, but did not improve postoperative morbidity and mortality.