EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Defect-Engineered Metal-Organic Frameworks.
PMID 26036179
Abstract
Defect engineering in metal-organic frameworks (MOFs) is an exciting concept for tailoring material properties, which opens up novel opportunities not only in sorption and catalysis, but also in controlling more challenging physical characteristics such as band gap as well as magnetic and electrical/conductive properties. It is challenging to structurally characterize the inherent or intentionally created defects of various types, and there have so far been few efforts to comprehensively discuss these issues. Based on selected reports spanning the last decades, this Review closes that gap by providing both a concise overview of defects in MOFs, or more broadly coordination network compounds (CNCs), including their classification and characterization, together with the (potential) applications of defective CNCs/MOFs. Moreover, we will highlight important aspects of "defect-engineering" concepts applied for CNCs, also in comparison with relevant solid materials such as zeolites or COFs. Finally, we discuss the future potential of defect-engineered CNCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B92303
4-tert-Butylcyclohexanone, 99%
C10H18O
157635
7,7,8,8-Tetracyanoquinodimethane, 98%
C12H4N4