EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Controllable synthesis of hollow mesoporous silica particles by a facile one-pot sol-gel method.
PMID 26036428
Abstract
A simple and facile one-pot sol-gel method is proposed for the fabrication of hollow mesoporous silica particles. Both the particle size and the shell thickness can be well controlled by moderately tuning some experimental parameters.