EMAIL THIS PAGE TO A FRIEND
Integrative cancer therapies
Curcumin Inhibits Invasiveness and Epithelial-Mesenchymal Transition in Oral Squamous Cell Carcinoma Through Reducing Matrix Metalloproteinase 2, 9 and Modulating p53-E-Cadherin Pathway.
PMID 26036622
Abstract
Epithelial-mesenchymal transition (EMT) and invasion play a critical role in cancer progression and metastasis. We have shown that low E-cadherin and high Twist expression are significantly correlated with prognostic survival prediction in oral squamous cell carcinoma (OSCC). This study aimed to determine the anti-invasive effect of curcumin on the expression of matrix metalloproteinases (MMPs) and of EMT regulators in OSCC. SCC-25 cells were treated with curcumin, and cell proliferation, invasion, and expression of MMPs and EMT regulators were assessed for cell viability by trypan blue exclusion, for invasion by Matrigel invasion chamber, and for EMT regulators and MMP changes in the levels of proteins by immunoblotting. Our data showed that curcumin treatment not only decreased the expression of MMP-2 and MMP-9 to inhibit invasiveness in oral cancer but also modulated the expression of EMT markers, such as Snail, Twist, and E-cadherin, and induced p53 expression that is crucial to EMT repression. Curcumin has the potential to become an adjunctive regimen for the prevention of cancer progression and metastasis in oral cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4065 Anti-Matrix Metalloproteinase-2 antibody produced in rabbit, ~1 mg/mL, IgG fraction of antiserum, buffered aqueous solution
M5177 Anti-Matrix Metalloproteinase-9, C-Terminal antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501891
Anti-MMP-2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501896
Anti-MMP-9, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA001939
Anti-MMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV20016
Anti-MMP2 antibody produced in rabbit, IgG fraction of antiserum
AMAB90805
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0539, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90804
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0538, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300247
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5G3, ascites fluid
SAB5300248
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5C3, ascites fluid
AMAB90806
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0542, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402274
Monoclonal Anti-MMP9 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution