EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Comparative proteomic analysis using 2DE-LC-MS/MS reveals the mechanism of Fuzhuan brick tea extract against hepatic fat accumulation in rats with nonalcoholic fatty liver disease.
PMID 26036873
Abstract
Fuzhuan brick tea has received increasing attention in recent years owing to its benefits for nonalcoholic fatty liver disease (NAFLD) and associated metabolic syndrome. For exploring the ameliorative mechanism, the liver proteomes from three groups of rats fed either a normal control diet (NCD), a high fat diet (HFD), or a HFD supplemented with high-dose Fuzhuan brick tea extract (FTE) (HFD + HFTE) were comprehensively compared by quantitative proteomics using 2DE-LC-MS/MS. This is the first study of the effects of tea aqueous extract on the liver proteome of rats suffering from metabolic syndrome. The results showed that 57 proteins displayed more than 1.5-fold differences in at least one of two comparisons of HFD versus NCD and HFD versus HFD + HFTE due to HFD feeding and FTE treatment, respectively. Of them, over 75% of proteins exhibited a similar tendency of expression in the two comparisons, meaning FTE was able to correct HFD effects on rat livers. By function analyses, an extensive list of proteins was involved in sugar and lipid metabolism. Compared with HFD-fed rats, the reduced lipogenesis and enhanced β-oxidation, tricarboxylic acid cycle and respiratory chain in HFD + HFTE-fed rats, which mainly contributed to ameliorate hepatic fat accumulation and associated NAFLD. Additionally, some putative drug targets were also revealed such as COX2, PGAM1, ACACB, FAS, and ECHS1.