EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Brg1 Controls the Expression of Pax7 to Promote Viability and Proliferation of Mouse Primary Myoblasts.
PMID 26036967
Abstract
Brg1 (Brahma-related gene 1) is a catalytic component of the evolutionarily conserved mammalian SWI/SNF ATP-dependent chromatin remodeling enzymes that disrupt histone-DNA contacts on the nucleosome. While the requirement for the SWI/SNF enzymes in cell differentiation has been extensively studied, its role in precursor cell proliferation and survival is not as well defined. Muscle satellite cells constitute the stem cell pool that sustains and regenerates myofibers in adult skeletal muscle. Here, we show that deletion of Brg1 in primary mouse myoblasts derived from muscle satellite cells cultured ex vivo leads to a cell proliferation defect and apoptosis. We determined that Brg1 regulates cell proliferation and survival by controlling chromatin remodeling and activating transcription at the Pax7 promoter, which is expressed during somite development and is required for controlling viability of the satellite cell population. Reintroduction of catalytically active Brg1 or of Pax7 into Brg1-deficient satellite cells rescued the apoptotic phenotype and restored proliferation. These data demonstrate that Brg1 functions as a positive regulator for cellular proliferation and survival of primary myoblasts. Therefore, the regulation of gene expression through Brg1-mediated chromatin remodeling is critical not just for skeletal muscle differentiation but for maintaining the myoblast population as well.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA027241
Anti-ECM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32736
Anti-PAX3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32445
Anti-PAX3 (AB2) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2500753
Anti-PAX3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502032
Anti-PAX3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV32446
Anti-PAX3 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA063659
Anti-PAX3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101801
Anti-PI3 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101802
Anti-PI3 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1409446
Anti-PI3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA017737
Anti-PI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200195
Monoclonal Anti-Brg1 antibody produced in rat, ~1.0 mg/mL, clone 5B7, purified immunoglobulin
WH0005077M5
Monoclonal Anti-PAX3 antibody produced in mouse, clone 4F4, purified immunoglobulin, buffered aqueous solution
SAB1404167
Monoclonal Anti-PAX3 antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
WH0005266M1
Monoclonal Anti-PI3 antibody produced in mouse, clone 3G9, purified immunoglobulin, buffered aqueous solution
SAB1409445
Monoclonal Anti-PI3 antibody produced in mouse, clone 2G20, purified immunoglobulin, buffered aqueous solution