EMAIL THIS PAGE TO A FRIEND
Journal of food protection
Evaluation of commercial kit based on loop-mediated isothermal amplification for rapid detection of low levels of uninjured and injured Salmonella on duck meat, bean sprouts, and fishballs in Singapore.
PMID 26038914
Abstract
The objective of this study was to evaluate performance of the commercial kit based on loop-mediated isothermal amplification (LAMP) in comparison with the International Organization for Standardization method for detecting uninjured and sublethally injured Salmonella cells artificially inoculated at levels of 10(0) and 10(1) CFU/25 g on raw duck wing, raw mung bean sprouts, and processed fishballs. Injured cells were prepared by a heat treatment for duck wings and fishball samples and a chlorine treatment for bean sprout samples. Additionally, a validation study was performed on naturally contaminated food samples sold in Singapore. A total of 110 samples of each commodity were analyzed in this study. Regardless of inoculum levels, the detection by the commercial LAMP kit showed 100% sensitivity and specificity for both inoculated and uninoculated samples compared with the International Organization for Standardization method, with the exception of bean sprout samples. Only 20% of bean sprout samples inoculated with 10(0) CFU/25 g injured Salmonella cells were positive by using the commercial LAMP-based kit. However, all negative samples became positive following a secondary enrichment in Rappaport-Vassiliadis medium with soy broth or after concentration by centrifugation. These results suggest that secondary enrichment or centrifugation should be considered as an additional step to increase the sensitivity of the commercial LAMP-based kit with low numbers of injured target cells in samples with high background microflora (such as mung bean sprouts). The validation study also showed that the commercial LAMP-based kit provided 91% sensitivity and 95% specificity for naturally contaminated samples. Thus, this study demonstrates that the commercial LAMP-based kit might be a cost-effective method, as this system could provide rapid, accurate detection of both uninjured and injured Salmonella cells on raw duck wings, raw mung bean sprouts, and processed fishballs in less than 26 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295132
Chlorine, ≥99.5%
Cl2