EMAIL THIS PAGE TO A FRIEND
Food chemistry
Separation and characterization of alpha-chain subunits from tilapia (Tilapia zillii) skin gelatin using ultrafiltration.
PMID 26041203
Abstract
Alpha-chain subunits were separated from tilapia skin gelatin using ultrafiltration, and the physicochemical properties of obtained subunits were investigated. As a result, α1-subunit and α2-subunit could be successfully separated by 100 kDa MWCO regenerated cellulose membranes and 150 kDa MWCO polyethersulfone membranes, respectively. Glycine was the most dominant amino acid in both α1-subunit and α2-subunit. However, the tyrosine content was higher in α2-subunit than in α1-subunit, resulting in strong absorption near 280 nm observed in the UV absorption spectrum. Based on the DSC analysis, it was found that the glass transition temperatures of gelatin, α1-subunit and α2-subunit were 136.48 °C, 126.77 °C and 119.43 °C, respectively. Moreover, the reduced viscosity and denaturation temperature of α1-subunit were higher than those of α2-subunit, and the reduced viscosity reached the highest when α-subunits were mixed with α1/α2 ratio of approximately 2, suggesting that α1-subunit plays a more important role in the thermostability of gelatin than α2-subunit.