EMAIL THIS PAGE TO A FRIEND
Journal of virology
Early Steps of Jaagsiekte Sheep Retrovirus-Mediated Cell Transformation Involve the Interaction between Env and the RALBP1 Cellular Protein.
PMID 26041289
Abstract
Ovine pulmonary adenocarcinoma is a naturally occurring lung cancer in sheep induced by the Jaagsiekte sheep retrovirus (JSRV). Its envelope glycoprotein (Env) carries oncogenic properties, and its expression is sufficient to induce in vitro cell transformation and in vivo lung adenocarcinoma. The identification of cellular partners of the JSRV envelope remains crucial for deciphering mechanisms leading to cell transformation. We initially identified RALBP1 (RalA binding protein 1; also known as RLIP76 or RIP), a cellular protein implicated in the ras pathway, as a partner of JSRV Env by yeast two-hybrid screening and confirmed formation of RALBP1/Env complexes in mammalian cells. Expression of the RALBP1 protein was repressed in tumoral lungs and in tumor-derived alveolar type II cells. Through its inhibition using specific small interfering RNA (siRNA), we showed that RALBP1 was involved in envelope-induced cell transformation and in modulation of the mTOR (mammalian target of rapamycin)/p70S6K pathway by the retroviral envelope. JSRV-induced lung adenocarcinoma is of importance for the sheep industry. While the envelope has been reported as the oncogenic determinant of the virus, the cellular proteins directly interacting with Env are still not known. Our report on the formation of RALBP/Env complexes and the role of this interaction in cell transformation opens up a new hypothesis for the dysregulation observed upon virus infection in sheep.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5747
Anti-CDC42 (23-35) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500462
Anti-CDC42 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2100382
Anti-CDC42 antibody produced in rabbit, affinity isolated antibody
SAB1411717
Anti-CDC42 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA046651
Anti-RALBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410141
Anti-RALBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105823
Anti-RALBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1408145
Anti-REPS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029961
Anti-REPS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0010928M2
Monoclonal Anti-RALBP1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1404880 Monoclonal Anti-RALBP1 antibody produced in mouse, clone 1C2-A10, purified immunoglobulin, buffered aqueous solution
SAB4100176
Monoclonal Anti-RALBP1 antibody produced in mouse, clone RALBP1-A192, culture supernatant
394831
N,O-Bis(trimethylsilyl)acetamide, for GC derivatization
C8H21NOSi2
128910
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
C8H21NOSi2
P6061
Phenylacetic acid, plant cell culture tested
C8H8O2
P16621
Phenylacetic acid, 99%
C8H8O2
W287814
Phenylacetic acid, natural, ≥99%
C8H8O2
W287806
Phenylacetic acid, ≥99%, FCC, FG
C8H8O2
R8781 Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
C51H79NO13
N3398
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
C7H12N2O4S