EMAIL THIS PAGE TO A FRIEND
Nature communications
Molecular architecture of native fibronectin fibrils.
PMID 26041410
Abstract
Fibronectin fibrils within the extracellular matrix play central roles in physiological and pathological processes, yet many structural details about their hierarchical and molecular assembly remain unknown. Here we combine site-specific protein labelling with single-molecule localization by stepwise photobleaching or direct stochastic optical reconstruction microscopy (dSTORM), and determine the relative positions of various labelled sites within native matrix fibrils. Single end-labelled fibronectin molecules in fibrils display an average end-to-end distance of ∼133 nm. Sampling of site-specific antibody epitopes along the thinnest fibrils (protofibrils) shows periodic punctate label patterns with ∼95 nm repeats and alternating N- and C-terminal regions. These measurements suggest an antiparallel 30-40 nm overlap between N-termini, suggesting that the first five type I modules bind type III modules of the adjacent molecule. Thicker fibres show random bundling of protofibrils without a well-defined line-up. This super-resolution microscopy approach can be applied to other fibrillar protein assemblies of unknown structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500974
Anti-Fibronectin 1, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW20021F Anti-Fibronectin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405825
Anti-FN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV41490
Anti-FN1 antibody produced in rabbit, affinity isolated antibody
HPA027066
Anti-FN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104810
Anti-FN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB3700280 Anti-goat IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700328 Anti-Goat IgG (H+L), F(ab) fragment- antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB4600361
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600394
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SCJ4600055 CF 680 succinimidyl ester, suitable for fluorescence, ≥90% (TLC)
M9768
Cysteamine, ~95%
C2H7NS
30070
Cysteamine, ≥98.0% (RT)
C2H7NS
G2133
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
F7387 Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid
F0916 Monoclonal Anti-Fibronectin antibody produced in mouse, clone IST-4, ascites fluid
F0791 Monoclonal Anti-Fibronectin antibody produced in mouse, clone IST-3, ascites fluid