EMAIL THIS PAGE TO A FRIEND
Oncotarget
Chronic chemotherapeutic stress promotes evolution of stemness and WNT/beta-catenin signaling in colorectal cancer cells: implications for clinical use of WNT-signaling inhibitors.
PMID 26041882
Abstract
Most solid tumors contain a subfraction of cells with stem/progenitor cell features. Stem cells are naturally chemoresistant suggesting that chronic chemotherapeutic stress may select for cells with increased "stemness". We carried out a comprehensive molecular and functional analysis of six independently selected colorectal cancer (CRC) cell lines with acquired resistance to three different chemotherapeutic agents derived from two distinct parental cell lines. Chronic drug exposure resulted in complex alterations of stem cell markers that could be classified into three categories: 1) one cell line, HT-29/5-FU, showed increased "stemness" and WNT-signaling, 2) three cell lines showed decreased expression of stem cell markers, decreased aldehyde dehydrogenase activity, attenuated WNT-signaling and lost the capacity to form colonospheres and 3) two cell lines displayed prominent expression of ABC transporters with a heterogeneous response for stem cell markers. While WNT-signaling could be attenuated in the HT-29/5-FU cells by the WNT-signaling inhibitors ICG-001 and PKF-118, this was not accompanied by any selective growth inhibitory effect suggesting that the cytotoxic activity of these compounds is not directly linked to WNT-signaling inhibition. We conclude that classical WNT-signaling inhibitors have toxic off-target activities that need to be addressed for clinical development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA010926
Anti-ALCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C2206
Anti-β-Catenin antibody produced in rabbit, whole antiserum
SAB1306488
ANTI-CD166(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22755 Anti-CD44 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405590
Anti-CD44 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005785
Anti-CD44 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300691
Anti-CD44 antibody produced in rabbit, affinity isolated antibody
SAB4300470
Anti-CTNNB1 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300471
Anti-CTNNB1 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300381
Anti-CTNNB1 (Ab-41/45) antibody produced in rabbit, affinity isolated antibody
SAB4300586
Anti-CTNNB1 (Ab-654) antibody produced in rabbit, affinity isolated antibody
AV14001
Anti-CTNNB1 (AB1) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2701829
Anti-ctnnb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701804
Anti-ctnnb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100502
Anti-CTNNB1 antibody produced in rabbit, affinity isolated antibody
HPA029159
Anti-CTNNB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA029160
Anti-CTNNB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA012530
Anti-LGR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107606
Anti-PROM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1402685
Monoclonal Anti-ALCAM antibody produced in mouse, clone 1H3, ascites fluid
SAB5300346
Monoclonal Anti-ALCAM antibody produced in mouse, clone 3F8B12, ascites fluid
SAB5300347 Monoclonal Anti-ALCAM antibody produced in mouse, clone 10F1G12, ascites fluid
C7082
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 6F9, ascites fluid
C7207
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid
C7923 Monoclonal Anti-CD44 antibody produced in mouse, clone A3D8, purified immunoglobulin, buffered aqueous solution
SAB1402714
Monoclonal Anti-CD44 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
SAB4700179 Monoclonal Anti-CD44 antibody produced in mouse, clone MEM-85, purified immunoglobulin, buffered aqueous solution
SAB4700184 Monoclonal Anti-CD44 antibody produced in mouse, clone MEM-263, purified immunoglobulin, buffered aqueous solution
SAB4700188 Monoclonal Anti-CD44 antibody produced in rat, clone IM7, purified immunoglobulin, buffered aqueous solution
WH0001499M2
Monoclonal Anti-CTNNB1 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution