EMAIL THIS PAGE TO A FRIEND
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group
Hyperthermia effects on Hsp27 and Hsp72 associations with mismatch repair (MMR) proteins and cisplatin toxicity in MMR-deficient/proficient colon cancer cell lines.
PMID 26043026
Abstract
Hyperthermia is used in combination with conventional anticancer agents to potentiate their cytotoxicity. One of its key events is the synthesis of heat shock proteins (HSPs), which are able to associate with components from DNA repair mechanisms. However, little is known about their relationship with the mismatch repair system (MMR). Our aim was to study the effects of hyperthermia on cisplatin (cPt) sensitivity and to determine whether MLH1 and MSH2 associate with Hsp27 and Hsp72 in MMR-deficient(-)/-proficient(+) cells. HCT116+ch2 (MMR-) and HCT116+ch3 (MMR+) cell lines were exposed to cPt with or without previous hyperthermia (42 °C, 1 h). Clonogenic survival assays, MTT, confocal immunofluorescence, immunoprecipitation, immunoblotting and flow cytometry were performed. Hyperthermia increased the cPt resistance in MMR- cells 1.42-fold. Immunofluorescence revealed that after cPt, Hsp27 and Hsp72 translocated to the nucleus and colocalisation coefficients between these proteins with MLH1 and MSH2 increased in MMR+ cells. Immunoprecipitation confirmed the interactions between HSPs and MMR proteins in control and treated cells. Hyperthermia pretreatment induced cell cycle arrest, increased p73 expression and potentiated cPt sensitivity in MMR+ cells. This is the first report showing in a MMR-/+ cellular model that MLH1 and MSH2 are client proteins of Hsp27 and Hsp72. Our study suggests that p73 might participate in the cellular response to hyperthermia and cPt in a MMR-dependent manner. Further functional studies will confirm whether HSPs cooperate with the MMR system in cPt-induced DNA damage response or whether these protein interactions are only the result of their chaperone functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401870
Anti-ARHGAP24 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014288 Anti-ARHGAP24 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33096
Anti-HSPA1A antibody produced in rabbit, IgG fraction of antiserum
SAB2700846
Anti-HSPA1A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701085
Anti-HSPA1A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107600
Anti-HSPA1A antibody produced in rabbit, affinity isolated antibody
SAB4300495
Anti-HSPB1 (Ab-78) antibody produced in rabbit, affinity isolated antibody
SAB4300496
Anti-HSPB1 (Ab-82) antibody produced in rabbit, affinity isolated antibody
AV09060
Anti-HSPB1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV30177
Anti-HSPB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA000497
Anti-HSPB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701965
Anti-HSPB1 antibody produced in rabbit
M8445
Anti-MLH1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
M8320
Anti-MLH1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4501881
Anti-MLH1 antibody produced in rabbit, affinity isolated antibody
SAB2104053
Anti-MLH1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
AV41350
Anti-MSH2 antibody produced in rabbit, IgG fraction of antiserum
SAB2101508
Anti-MSH2 antibody produced in rabbit, affinity isolated antibody
SAB2700170
Anti-MSH2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501923
Anti-MSH2 antibody produced in rabbit, affinity isolated antibody
WH0003304M2
Monoclonal Anti-HSPA1B antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1403949
Monoclonal Anti-HSPA1B antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
WH0004292M2
Monoclonal Anti-MLH1 antibody produced in mouse, clone M1, purified immunoglobulin, buffered aqueous solution
SAB5300249 Monoclonal Anti-MLH1 antibody produced in mouse, clone 4C9C7, ascites fluid
WH0004436M2
Monoclonal Anti-MSH2 antibody produced in mouse, clone 4B2, purified immunoglobulin, buffered aqueous solution
SAB1404083
Monoclonal Anti-MSH2, (C-terminal) antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
PLA0004
Rabbit anti-MLH1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0106
Rabbit anti-MSH2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0040
Rabbit anti-p73 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.