EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Research on the interaction of hydrogen-bond acidic polymer sensitive sensor materials with chemical warfare agents simulants by inverse gas chromatography.
PMID 26043177
Abstract
Hydrogen-bond acidic polymers are important high affinity materials sensitive to organophosphates in the chemical warfare agent sensor detection process. Interactions between the sensor sensitive materials and chemical warfare agent simulants were studied by inverse gas chromatography. Hydrogen bonded acidic polymers, i.e., BSP3, were prepared for micro-packed columns to examine the interaction. DMMP (a nerve gas simulant) and 2-CEES (a blister agent simulant) were used as probes. Chemical and physical parameters such as heats of absorption and Henry constants of the polymers to DMMP and 2-CEES were determined by inverse gas chromatography. Details concerning absorption performance are also discussed in this paper.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
448621
1,1,3,3,5,5-Hexamethyltrisiloxane, 95%
C6H20O2Si3
242640
2-Chloroethyl ethyl sulfide, 97%
C4H9ClS