EMAIL THIS PAGE TO A FRIEND
mBio
Development of a Model System for Tick-Borne Flavivirus Persistence in HEK 293T Cells.
PMID 26045539
Abstract
We devised a model system to study persistent infection by the tick-borne flavivirus Langat virus (LGTV) in 293T cells. Infection with a molecularly cloned LGTV strain produced an acute lytic crisis that left few surviving cells. The culture was repopulated by cells that were ~90% positive for LGTV E protein, thus initiating a persistent infection that was maintained for at least 35 weeks without additional lytic crises. Staining of cells for viral proteins and ultrastructural analysis revealed only minor differences from the acute phase of infection. Infectious LGTV decreased markedly over the study period, but the number of viral genomes remained relatively constant, suggesting the development of defective interfering particles (DIPs). Viral genome changes were investigated by RNA deep sequencing. At the initiation of persistent infection, levels of DIPs were below the limit of detection at a coverage depth of 11,288-fold, implying that DIPs are not required for initiation of persistence. However, after 15 passages, DIPs constituted approximately 34% of the total LGTV population (coverage of 1,293-fold). Furthermore, at this point, one specific DIP population predominated in which nucleotides 1058 to 2881 had been deleted. This defective genome specified an intact polyprotein that coded for a truncated fusion protein containing 28 N-terminal residues of E and 134 C-terminal residues of NS1. Such a fusion protein has not previously been described, and a possible function in persistent infection is uncertain. DIPs are not required for the initiation of persistent LGTV infection but may play a role in the maintenance of viral persistence. Tick-borne flaviviruses are significant infectious agents that cause serious disease and death in humans worldwide. Infections are characterized by severe neurological symptoms, such as meningitis and encephalitis. A high percentage of people who get infected and recuperate from the acute phase of infection continue to suffer from chronic debilitating neurological sequelae, most likely as a result of nervous tissue damage, viral persistence, or both. However, little is known about mechanisms of viral persistence. Therefore, we undertook studies to investigate the persistence of Langat virus, a member of the tick-borne flavivirus group, in a mammalian cell line. Using next-generation sequencing, we determined that defective viral genomes do not play a role in the initiation of persistence, but their occurrence seems to be nonstochastic and could play a role in the maintenance of viral persistence via the expression of a novel envelope-NS1 fusion protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9673
1-Bromo-3-chloropropane, for isolation of RNA
C3H6BrCl
B62404
1-Bromo-3-chloropropane, 99%
C3H6BrCl
SAB1304456
ANTI-PDIA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501845
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501851
Anti-PDIA2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051692
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053492
Anti-PDIA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405897
Anti-PDIA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002645
Anti-PDIA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003230
Anti-PDIA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107799
Anti-PDIA3 antibody produced in rabbit, affinity isolated antibody
SAB1410582
Anti-PDIA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution