EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Preparation and characterization of nano-sized hydroxyapatite/alginate/chitosan composite scaffolds for bone tissue engineering.
PMID 26046263
Abstract
The aim of this study was to develop chitosan composite scaffolds with high strength and controlled pore structures by homogenously dispersed nano-sized hydroxyapatite (nano-HAp) powders. In the fabrication of composite scaffolds, nano-HAp powders distributed in an alginate (AG) solution with a pH higher than 10 were mixed with a chitosan (CS) solution and then freeze dried. While the HAp content increased up to 70 wt.%, the compressive strength and the elastic modulus of the composite scaffolds significantly increased from 0.27 MPa and 4.42 MPa to 0.68 MPa and 13.35 MPa, respectively. Higher content of the HAp also helped develop more differentiation and mineralization of the MC3T3-E1 cells on the composite scaffolds. The uniform pore structure and the excellent mechanical properties of the HAp/CS composite scaffolds likely resulted from the use of the AG solution at pH 10 as a dispersant for the nano-HAp powders.