PloS one
Gastrointestinal Fibroblasts Have Specialized, Diverse Transcriptional Phenotypes: A Comprehensive Gene Expression Analysis of Human Fibroblasts.
PMID 26046848
Abstract
Fibroblasts are the principal stromal cells that exist in whole organs and play vital roles in many biological processes. Although the functional diversity of fibroblasts has been estimated, a comprehensive analysis of fibroblasts from the whole body has not been performed and their transcriptional diversity has not been sufficiently explored. The aim of this study was to elucidate the transcriptional diversity of human fibroblasts within the whole body. Global gene expression analysis was performed on 63 human primary fibroblasts from 13 organs. Of these, 32 fibroblasts from gastrointestinal organs (gastrointestinal fibroblasts: GIFs) were obtained from a pair of 2 anatomical sites: the submucosal layer (submucosal fibroblasts: SMFs) and the subperitoneal layer (subperitoneal fibroblasts: SPFs). Using hierarchical clustering analysis, we elucidated identifiable subgroups of fibroblasts and analyzed the transcriptional character of each subgroup. In unsupervised clustering, 2 major clusters that separate GIFs and non-GIFs were observed. Organ- and anatomical site-dependent clusters within GIFs were also observed. The signature genes that discriminated GIFs from non-GIFs, SMFs from SPFs, and the fibroblasts of one organ from another organ consisted of genes associated with transcriptional regulation, signaling ligands, and extracellular matrix remodeling. GIFs are characteristic fibroblasts with specific gene expressions from transcriptional regulation, signaling ligands, and extracellular matrix remodeling related genes. In addition, the anatomical site- and organ-dependent diversity of GIFs was also discovered. These features of GIFs contribute to their specific physiological function and homeostatic maintenance, and create a functional diversity of the gastrointestinal tract.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701079 Anti-Mouse IgG (H+L) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701163 Anti-Mouse IgG (H+L), F(ab) fragment antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700999 Anti-Mouse IgG F(ab′)2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701000 Anti-Mouse IgG F(ab′)2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV31396
Anti-MSX1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV37094
Anti-MSX1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500650 Anti-MSX1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412289 ANTI-MSX1 antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
M0944 Anti-Msx1 antibody produced in rabbit, affinity isolated antibody, ammonium sulfate suspension
M6198
Anti-MSX1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503083
Anti-Vimentin, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0004487M3 Monoclonal Anti-MSX1 antibody produced in mouse, clone 2B12, purified immunoglobulin, buffered aqueous solution
SAB1404087
Monoclonal Anti-MSX1 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB5300418
Monoclonal Anti-MSX1 antibody produced in mouse, clone 5D11, ascites fluid