EMAIL THIS PAGE TO A FRIEND
Marine drugs
Fucoidan Suppresses Hypoxia-Induced Lymphangiogenesis and Lymphatic Metastasis in Mouse Hepatocarcinoma.
PMID 26047481
Abstract
Metastasis, the greatest clinical challenge associated with cancer, is closely connected to multiple biological processes, including invasion and adhesion. The hypoxic environment in tumors is an important factor that causes tumor metastasis by activating HIF-1α. Fucoidan, extracted from brown algae, is a sulfated polysaccharide and, as a novel marine biological material, has been used to treat various disorders in China, Korea, Japan and other countries. In the present study, we demonstrated that fucoidan derived from Undaria pinnatifida sporophylls significantly inhibits the hypoxia-induced expression, nuclear translocation and activity of HIF-1α, the synthesis and secretion of VEGF-C and HGF, cell invasion and lymphatic metastasis in a mouse hepatocarcinoma Hca-F cell line. Fucoidan also suppressed lymphangiogenesis in vitro and in vivo. In addition, accompanied by a reduction in the HIF-1α nuclear translocation and activity, fucoidan significantly reduced the levels of p-PI3K, p-Akt, p-mTOR, p-ERK, NF-κB, MMP-2 and MMP-9, but increased TIMP-1 levels. These results indicate strongly that the anti-metastasis and anti-lymphangiogenesis activities of fucoidan are mediated by suppressing HIF-1α/VEGF-C, which attenuates the PI3K/Akt/mTOR signaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300609
Anti-FLT4 antibody produced in rabbit, affinity isolated antibody
M4065 Anti-Matrix Metalloproteinase-2 antibody produced in rabbit, ~1 mg/mL, IgG fraction of antiserum, buffered aqueous solution
M9570 Anti-Matrix Metalloproteinase-9 antibody produced in goat, affinity isolated antibody
M5177 Anti-Matrix Metalloproteinase-9, C-Terminal antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501891
Anti-MMP-2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501896
Anti-MMP-9, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA001939
Anti-MMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV20016
Anti-MMP2 antibody produced in rabbit, IgG fraction of antiserum
HPA001238
Anti-MMP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33090
Anti-MMP9 antibody produced in rabbit, IgG fraction of antiserum
SAB1411757
Anti-VEGFC antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1306762
ANTI-VEGFC antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1306761
ANTI-VEGFC antibody produced in mouse, crude ascites, buffered aqueous solution
409332
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Cl2Co
449776
Cobalt(II) chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Co
WH0002324M1
Monoclonal Anti-FLT4 antibody produced in mouse, clone 5B6, purified immunoglobulin, buffered aqueous solution
SAB1403833
Monoclonal Anti-FLT4 antibody produced in mouse, clone 5B5, purified immunoglobulin, buffered aqueous solution
SAB1409022
Monoclonal Anti-FLT4 antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution
AMAB90805
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0539, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90804
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0538, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300247
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5G3, ascites fluid
SAB5300248
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5C3, ascites fluid
AMAB90806
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0542, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402274
Monoclonal Anti-MMP9 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1305566
MONOCLONAL ANTI-VEGFR3 antibody produced in mouse, clone 818CT12.1.1, IgG fraction of antiserum, buffered aqueous solution