EMAIL THIS PAGE TO A FRIEND
International journal of oral science
Bone morphogenetic protein 2-induced human dental pulp cell differentiation involves p38 mitogen-activated protein kinase-activated canonical WNT pathway.
PMID 26047580
Abstract
Both bone morphogenetic protein 2 (BMP2) and the wingless-type MMTV integration site (WNT)/β-catenin signalling pathway play important roles in odontoblast differentiation and dentinogenesis. Cross-talk between BMP2 and WNT/β-catenin in osteoblast differentiation and bone formation has been identified. However, the roles and mechanisms of the canonical WNT pathway in the regulation of BMP2 in dental pulp injury and repair remain largely unknown. Here, we demonstrate that BMP2 promotes the differentiation of human dental pulp cells (HDPCs) by activating WNT/β-catenin signalling, which is further mediated by p38 mitogen-activated protein kinase (MAPK) in vitro. BMP2 stimulation upregulated the expression of β-catenin in HDPCs, which was abolished by SB203580 but not by Noggin or LDN193189. Furthermore, BMP2 enhanced cell differentiation, which was not fully inhibited by Noggin or LDN193189. Instead, SB203580 partially blocked BMP2-induced β-catenin expression and cell differentiation. Taken together, these data suggest a possible mechanism by which the elevation of β-catenin resulting from BMP2 stimulation is mediated by the p38 MAPK pathway, which sheds light on the molecular mechanisms of BMP2-mediated pulp reparative dentin formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4504206
Anti-phospho-Smad1 (pSer187) antibody produced in rabbit, affinity isolated antibody
SAB4503779
Anti-phospho-Smad1 (pSer465) antibody produced in rabbit, affinity isolated antibody
SAB4300250
Anti-phospho-SMAD1 (pSer465) antibody produced in rabbit, affinity isolated antibody
SAB4300561
Anti-SMAD1 (Ab-465) antibody produced in rabbit, affinity isolated antibody
AV38693
Anti-SMAD1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV38692
Anti-SMAD1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB4200377
Anti-SMAD1 (Internal region) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1412181
ANTI-SMAD1 antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB1412594
ANTI-SMAD1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
SAB1412595
ANTI-SMAD1 antibody produced in mouse, clone 4C12, purified immunoglobulin, buffered aqueous solution
SAB1412656
ANTI-SMAD1 antibody produced in mouse, clone 1B8, purified immunoglobulin, buffered aqueous solution
AV32003
Anti-SMAD1 antibody produced in rabbit, affinity isolated antibody
SAB4200378
Anti-SMAD1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4501802
Anti-SMAD1 antibody produced in rabbit, affinity isolated antibody
SAB4501801
Anti-SMAD1, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0004086M2
Monoclonal Anti-SMAD1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1404035
Monoclonal Anti-SMAD1 antibody produced in mouse, clone 2E9, purified immunoglobulin, buffered aqueous solution
X3004
XAV939, ≥98% (HPLC)
C14H11F3N2OS