EMAIL THIS PAGE TO A FRIEND
International urology and nephrology
Correlation between arterial stiffness and inflammatory markers in autosomal dominant polycystic kidney disease patients with preserved renal function.
PMID 26047596
Abstract
To evaluate the association between arterial stiffness and inflammatory markers including C-reactive protein (CRP), pentraxin 3 (PTX3) and neutrophil-to-lymphocyte ratio (NLR) in autosomal dominant polycystic kidney disease (ADPKD) patients with preserved renal function. A total of 52 ADPKD patients [mean (SD) age 38.2 (12.8) years, 69.2 % were females] with preserved renal function and 25 healthy volunteers [mean (SD) age 35.5 (6.5) years, 48.0 % were females] were included. Data on patient characteristics, blood biochemistry, inflammatory markers [PTX3 (pg/mL), CRP (mg/dL) and NLR] and arterial stiffness [large artery elasticity index (LAEI) (mL/mmHg × 10) and small artery elasticity index (SAEI) (mL/mmHg × 100)] were recorded in patient and control groups. Correlation between inflammatory markers and arterial stiffness parameters was analysed in patients. Overall, 42.3 % of ADPKD patients were hypertensive and 44.4 % were receiving renin-angiotensin-aldosterone system (RAAS) blockade therapy. Median levels for PTX3 [442.0 (20.0-4140.0) pg/mL vs. 220.5 (14.7-393.0) pg/mL, p < 0.001] and SAEI [4.90 (1.60-11.80) mL/mmHg × 100 vs. 6.45 (2.80-15.70) mL/mmHg × 10, p = 0.013] were significantly higher in ADPKD patients than in controls. PTX3 and CRP were not correlated with arterial elasticity, while NLR was significantly correlated with LAEI negatively (Rho = -0.278, p = 0.042). In conclusion, our findings revealed increased PTX3 levels and reduced SAEI in patients as compared with controls, while no correlation between inflammatory markers studied and the small artery elasticity.