EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Ascorbic acid promotes the direct conversion of mouse fibroblasts into beating cardiomyocytes.
PMID 26047705
Abstract
Recent advances in the direct conversion of fibroblasts to cardiomyocytes suggest this process as a novel promising approach for cardiac cell-based therapies. Here, by screening the effects of 10 candidate small molecules along with transient overexpression of Yamanaka factors, we show ascorbic acid (AA), also known as vitamin C, enhances reprogramming of mouse fibroblasts into beating cardiomyocytes. Immunostaining and gene expression analyses for pluripotency and cardiac lineage markers confirmed beating patches were derived from non-cardiac lineage cells without passing through a pluripotent intermediate. Further analysis revealed that AA also increased the size of the beating areas and the number of cardiac progenitors. Immunostaining for cardiac markers, as well as electrophysiological analysis confirmed the functionality of directly converted cardiomyocytes. These results illustrate the importance of AA in direct conversion of fibroblasts to cardiomyocytes and may open new insights into future biomedical applications for induced cardiomyocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5581
3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene, ≥95%
C4H13N5O2
X3004
XAV939, ≥98% (HPLC)
C14H11F3N2OS