EMAIL THIS PAGE TO A FRIEND
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.]
Metabolomic profiles illuminate the efficacy of Chinese herbal Da-Cheng-Qi decoction on acute pancreatitis in rats.
PMID 26048200
Abstract
Chinese herbal drug Da-Cheng-Qi decoction (DCQD) has been widely used for decades to treat acute pancreatitis (AP). Previous trials are mostly designed to state the potential mechanisms of the therapeutic effects rather than to detect its whole effect on metabolism. This study aimed to investigate the efficacy of DCQD on metabolism in AP. Twenty-two male adult Sprague-Dawley rats were randomized into three groups. AP was induced by retrograde ductal infusion of 3.5% sodium taurocholate solution in DCQD and AP group, while 0.9% saline solution was used in sham operation (SO) group. Blood samples were obtained 12xa0h after drug administration and a 600xa0MHz superconducting Nuclear Magnetic Resonance (NMR) spectrometer was used to detected plasma metabolites. Principal Components Analysis (PCA) and Partial Least Squares-Discriminant Analysis after Orthogonal Signal Correction (OSC-PLS-DA) were applied to analyze the Longitudinal Eddy-delay (LED) and Carr-Purcell-Meiboom-Gill (CPMG) spectra. Differences in concentrations of metabolites among the three groups were detected by OSC-PLS-DA of 1HNMR spectra (both LED and CPMG). Compared with SO group, DCQD group had higher levels of plasma glycerol, glutamic acid, low density lipoprotein (LDL), saturated fatty acid (FA) and lower levels of alanine and glutamine, while the metabolic changes were reversed in the AP group. Our results demonstrated that DCQD was capable of altering the changed concentrations of metabolites in rats with AP and 1HNMR-based metabolomic approach provided a new methodological cue for systematically investigating the efficacies and mechanisms of DCQD in treating AP.