EMAIL THIS PAGE TO A FRIEND
Journal of hematology & oncology
Targeting Wnt pathway in mantle cell lymphoma-initiating cells.
PMID 26048374
Abstract
Mantle cell lymphoma (MCL) is an aggressive and incurable form of non-Hodgkin's lymphoma. Despite initial intense chemotherapy, up to 50% of cases of MCL relapse often in a chemoresistant form. We hypothesized that the recently identified MCL-initiating cells (MCL-ICs) are the main reason for relapse and chemoresistance of MCL. Cancer stem cell-related pathways such as Wnt could be responsible for their maintenance and survival. We isolated MCL-ICs from primary MCL cells on the basis of a defined marker expression pattern (CD34-CD3-CD45+CD19-) and investigated Wnt pathway expression. We also tested the potential of Wnt pathway inhibitors in elimination of MCL-ICs. We showed that MCL-ICs are resistant to genotoxic agents vincristine, doxorubicin, and the newly approved Burton tyrosine kinase (BTK) inhibitor ibrutinib. We confirmed the differential up-regulation of Wnt pathway in MCL-ICs. Indeed, MCL-ICs were particularly sensitive to Wnt pathway inhibitors. Targeting β-catenin-TCF4 interaction with CCT036477, iCRT14, or PKF118-310 preferentially eliminated the MCL-ICs. Our results suggest that Wnt signaling is critical for the maintenance and survival of MCL-ICs, and effective MCL therapy should aim to eliminate MCL-ICs through Wnt signaling inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0151
CCT036477, ≥98% (HPLC)
C21H18ClN3
SML0203
iCRT14, ≥98% (HPLC)
C21H17N3O2S
X3004
XAV939, ≥98% (HPLC)
C14H11F3N2OS