EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Rosiglitazone suppresses HIV-1 Tat-induced vascular inflammation via Akt signaling.
PMID 26048716
Abstract
Peroxisome proliferator-activated receptor gamma (PPARƔ) contributes to human immunodeficiency virus (HIV)-1-induced dysfunction of brain endothelial cells. The aim of the present study was to evaluate the protection mechanism of PPARƔ against Tat-induced responses of adhesion molecules. We measured the protein expressions of intercellular adhesion molecule (ICAM)-1 and vascular cell adhesion molecule (VCAM)-1 in human brain microvascular endothelial cells (hCMEC/D3) and C57BL/6J mouse brain microvessels with Western blotting and immunofluorescent labeling. The mRNA levels of ICAM-1 and VCAM-1 were determined by real-time reverse-transcriptase polymerase chain reaction. HIV-1 Tat induced overexpression of ICAM-1 but not VCAM-1 in both hCMEC/D3 and brain microvessels, this response was attenuated by treatment with the PPARƔ agonist rosiglitazone. Tat-mediated upregulation of ICAM-1 and VCAM-1 levels were abolished by the addition of PPARƔ antagonist GW9662 and the Akt inhibitor KP3721, indicating that Akt signaling is involved in the PPARƔ-mediated protection of Tat-induced adhesion molecule upregulation. These results show that Akt signaling plays a key role in PPARƔ's vascular inflammatory effects that contribute to blood-brain barrier damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
642355
2-Chloro-5-nitrobenzanilide, 97%
C13H9ClN2O3
M6191
GW9662, >98% (HPLC)
C13H9ClN2O3